Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104462.4 | NP_175981.2 | 5 - 6 | 1962 | 69 | |
GenBank mRNA | AK227116.1 | - | 5 - 6 | 1704 | 69 | |
AY064040.1 | - | 5 - 6 | 1703 | 69 | ||
KJ138961.1 | - | 5 - 6 | 1650 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |