Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104516.1 | NP_176033.1 | 6 - 7 | 1198 | 74 | |
GenBank mRNA | BT026359.1 | - | 6 - 7 | 1198 | 74 | |
DQ446366.1 | - | 6 - 7 | 1198 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |