Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104521.3 | NP_176038.1 | 1 | 1141 | 120 | |
GenBank mRNA | AY099758.1 | - | 1 | 1032 | 120 | |
AY128890.1 | - | 1 | 953 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |