Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104523.4 | NP_176040.1 | 6 - 7 | 965 | 64 | |
GenBank mRNA | AF378880.1 | - | 6 - 7 | 704 | 64 | |
AK221762.1 | - | 6 - 7 | 715 | 64 | ||
AY045596.1 | - | 6 - 7 | 681 | 64 | ||
AY052739.1 | - | 5 - 6 | 628 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |