Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GolS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104537.3 | NP_176053.1 | 3 - 4 | 889 | 88 | |
GenBank mRNA | AB062849.1 | - | 3 - 4 | 885 | 88 | |
AY050410.1 | - | 3 - 4 | 889 | 88 | ||
AY058238.1 | - | 3 - 4 | 806 | 88 | ||
KJ138960.1 | - | 3 - 4 | 806 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |