Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198327.1 | NP_001185256.1 | 11 - 12 | 1437 | 104 | |
NM_001333808.1 | NP_001319262.1 | 10 - 11 | 1324 | 104 | ||
NM_001333809.1 | NP_001320412.1 | 11 - 12 | 1730 | 104 | ||
NM_104580.5 | NP_176096.1 | 11 - 12 | 1667 | 104 | ||
GenBank mRNA | BT002018.1 | - | 11 - 12 | 1440 | 104 | |
BT008861.1 | - | 9 - 10 | 989 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |