Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198330.1 | NP_001185259.1 | 2 - 3 | 286 | 126 | |
NM_001333816.1 | NP_001320703.1 | 2 - 3 | 410 | 126 | ||
NM_001333817.1 | NP_001320705.1 | 1 - 2 | 366 | 126 | ||
NM_104599.5 | NP_176114.2 | 2 - 3 | 410 | 126 | ||
NM_179492.4 | NP_849823.1 | 2 - 3 | 348 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |