Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198331.1 | NP_001185260.1 | 6 - 7 | 968 | 87 | |
NM_001333819.1 | NP_001322351.1 | 3 - 4 | 908 | 87 | ||
NM_001333820.1 | NP_001322352.1 | 3 - 4 | 538 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |