Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198333.1 | NP_001185262.1 | 15 - 16 | 6546 | 98 | |
NM_104605.4 | NP_176121.3 | 14 - 15 | 6521 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |