Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198333.1 | NP_001185262.1 | 5 - 6 | 1179 | 79 | |
NM_104605.4 | NP_176121.3 | 5 - 6 | 1298 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Arabidopsis thaliana Probe spans exons |