Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104609.4 | NP_176125.1 | 2 - 3 | 856 | 81 | |
GenBank mRNA | AY072223.1 | - | 2 - 3 | 654 | 81 | |
AY096600.1 | - | 2 - 3 | 505 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |