Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333829.1 | NP_001323263.1 | 4 - 5 | 918 | 91 | |
NM_104617.4 | NP_176133.1 | 4 - 5 | 615 | 91 | ||
GenBank mRNA | AB008015.1 | - | 4 - 5 | 597 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |