Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104654.5 | NP_176170.1 | 17 - 18 | 2302 | 69 | |
GenBank mRNA | AB028467.1 | - | 17 - 18 | 2204 | 69 | |
AY049230.1 | - | 17 - 18 | 2220 | 69 | ||
BT000772.1 | - | 17 - 18 | 2225 | 69 | ||
BT009695.1 | - | 17 - 18 | 2031 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |