Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104675.3 | NP_176191.1 | 16 - 17 | 1911 | 79 | |
GenBank mRNA | AY091777.1 | - | 16 - 17 | 1888 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |