Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198338.1 | NP_001185267.1 | 6 - 7 | 1042 | 98 | |
NM_001198339.2 | NP_001185268.1 | 6 - 7 | 1228 | 98 | ||
NM_001198340.2 | NP_001185269.1 | 6 - 7 | 1211 | 98 | ||
NM_104681.6 | NP_176197.3 | 6 - 7 | 1228 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |