Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E1 ALPHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333870.1 | NP_001321393.1 | 3 - 4 | 917 | 64 | |
NM_104682.3 | NP_176198.1 | 3 - 4 | 1079 | 64 | ||
GenBank mRNA | AY087667.1 | - | 3 - 4 | 904 | 64 | |
BT000974.1 | - | 3 - 4 | 844 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |