Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333876.1 | NP_001322340.1 | 4 - 5 | 340 | 131 | |
NM_104701.3 | NP_176217.2 | 4 - 5 | 317 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |