Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104735.4 | NP_176251.1 | 16 - 17 | 2506 | 88 | |
GenBank mRNA | BT000878.1 | - | 16 - 17 | 2307 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |