Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124046.2 | NP_001117518.1 | 9 - 10 | 1304 | 106 | |
NM_104743.3 | NP_176259.2 | 9 - 10 | 1296 | 106 | ||
GenBank mRNA | AK227267.1 | - | 9 - 10 | 1227 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |