Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRPAC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333919.1 | NP_001320360.1 | 8 - 9 | 1291 | 77 | |
NM_104766.5 | NP_176282.1 | 9 - 10 | 1185 | 77 | ||
NM_179502.4 | NP_849833.1 | 9 - 10 | 1174 | 77 | ||
NM_202328.3 | NP_974057.1 | 7 - 8 | 1303 | 77 | ||
GenBank mRNA | AK226775.1 | - | 9 - 10 | 1139 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |