Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333920.1 | NP_001322222.1 | 16 - 17 | 2283 | 75 | |
NM_001333921.1 | NP_001322221.1 | 10 - 11 | 1958 | 75 | ||
NM_104767.4 | NP_176283.1 | 18 - 19 | 2405 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |