Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198352.1 | NP_001185281.1 | 12 - 13 | 1254 | 74 | |
NM_001198354.1 | NP_001185283.1 | 12 - 13 | 1256 | 74 | ||
NM_001333983.1 | NP_001322619.1 | 12 - 13 | 1474 | 74 | ||
NM_001333988.1 | NP_001322614.1 | 11 - 12 | 1591 | 74 | ||
NM_104836.2 | NP_176350.1 | 12 - 13 | 1498 | 74 | ||
GenBank mRNA | AF369567.2 | - | 12 - 13 | 1253 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |