Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104850.2 | NP_176361.2 | 2 - 3 | 444 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |