Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104854.4 | NP_176365.1 | 5 - 6 | 914 | 85 | |
GenBank mRNA | DQ446384.1 | - | 5 - 6 | 868 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |