Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAS III | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036144.2 | NP_001031221.1 | 8 - 9 | 1264 | 77 | |
NM_104942.4 | NP_176452.1 | 7 - 8 | 1349 | 77 | ||
GenBank mRNA | AY063804.1 | - | 7 - 8 | 1209 | 77 | |
AY091275.1 | - | 7 - 8 | 993 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |