Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104972.3 | NP_176482.1 | 1 - 2 | 108 | 88 | |
GenBank mRNA | AY250836.1 | - | 1 - 2 | 86 | 88 | |
AY376734.1 | - | 1 - 2 | 86 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |