Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104977.3 | NP_564805.1 | 1 - 2 | 584 | 105 | |
GenBank mRNA | AF308451.1 | - | 1 - 2 | 359 | 105 | |
BT026059.1 | - | 1 - 2 | 283 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |