Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRS/ER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104978.5 | NP_564806.1 | 1 - 2 | 746 | 118 | |
GenBank mRNA | AF332445.1 | - | 1 - 2 | 447 | 118 | |
AY513232.1 | - | 1 - 2 | 447 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |