Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198369.1 | NP_001185298.1 | 11 - 12 | 3008 | 118 | |
NM_001334061.1 | NP_001320709.1 | 10 - 11 | 3198 | 118 | ||
NM_001334062.1 | NP_001320708.1 | 11 - 12 | 3008 | 118 | ||
NM_001334063.1 | NP_001320710.1 | 2 - 3 | 2095 | 118 | ||
NM_104980.4 | NP_176490.2 | 10 - 11 | 3173 | 118 | ||
GenBank mRNA | AY826515.1 | - | 9 - 10 | 2899 | 118 | |
DQ020657.1 | - | 10 - 11 | 3173 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |