Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125496.2 | NP_001118968.1 | 13 - 14 | 5323 | 88 | |
NM_001340762.1 | NP_001329652.1 | 13 - 14 | 5433 | 88 | ||
NM_001340763.1 | NP_001329653.1 | 12 - 13 | 5552 | 88 | ||
NM_001340765.1 | NP_001329655.1 | 13 - 14 | 5385 | 88 | ||
NM_001340767.1 | NP_001329657.1 | 13 - 14 | 5398 | 88 | ||
NM_117272.3 | NP_192939.2 | 13 - 14 | 5323 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |