Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117278.4 | NP_192945.2 | 1 - 2 | 739 | 93 | |
GenBank mRNA | AB284367.1 | - | 1 - 2 | 473 | 93 | |
AY070738.1 | - | 1 - 2 | 720 | 93 | ||
AY149932.1 | - | 1 - 2 | 473 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |