Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117317.4 | NP_192984.1 | 1 | 608 | 186 | |
GenBank mRNA | AY093032.1 | - | 1 | 566 | 186 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |