Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129010.4 | NP_181004.2 | 2 - 3 | 376 | 83 | |
GenBank mRNA | AY102549.1 | - | 2 - 3 | 376 | 83 | |
AY954813.1 | - | 2 - 3 | 275 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |