Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129010.4 | NP_181004.2 | 4 - 5 | 768 | 105 | |
GenBank mRNA | AY102549.1 | - | 4 - 5 | 768 | 105 | |
AY954813.1 | - | 4 - 5 | 667 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |