Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336523.1 | NP_001324697.1 | 25 - 26 | 3950 | 92 | |
NM_129022.8 | NP_181015.7 | 25 - 26 | 3761 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |