Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129025.4 | NP_181018.1 | 8 - 9 | 1487 | 97 | |
GenBank mRNA | AJ441291.1 | - | 8 - 9 | 1125 | 97 | |
AY099791.1 | - | 8 - 9 | 1403 | 97 | ||
BT000335.1 | - | 8 - 9 | 1125 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |