Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129040.4 | NP_565794.1 | 1 | 458 | 150 | |
GenBank mRNA | AY086924.1 | - | 1 | 435 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |