Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KUP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036407.2 | NP_001031484.1 | 8 - 9 | 2131 | 137 | |
NM_001336538.1 | NP_001324455.1 | 7 - 8 | 2803 | 137 | ||
NM_001336539.1 | NP_001324456.1 | 8 - 9 | 2141 | 137 | ||
NM_129058.5 | NP_181051.1 | 8 - 9 | 2256 | 137 | ||
GenBank mRNA | AK227188.1 | - | 8 - 9 | 2020 | 137 | |
AY099600.1 | - | 8 - 9 | 2016 | 137 | ||
BT002147.1 | - | 7 - 8 | 1881 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |