Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPSN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129072.4 | NP_565800.1 | 5 - 6 | 942 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |