Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334122.1 | NP_001319310.1 | 11 - 12 | 2684 | 84 | |
NM_105047.2 | NP_176557.1 | 10 - 11 | 2234 | 84 | ||
GenBank mRNA | AY357947.1 | - | 10 - 11 | 2234 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |