Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEH1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105109.4 | NP_564830.1 | 2 - 3 | 441 | 114 | |
GenBank mRNA | AF370222.1 | - | 2 - 3 | 428 | 114 | |
AY059116.1 | - | 2 - 3 | 371 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |