Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BDG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334171.1 | NP_001323396.1 | 1 - 2 | 728 | 87 | |
NM_001334172.1 | NP_001323395.1 | 1 - 2 | 686 | 87 | ||
NM_105142.4 | NP_564837.1 | 1 - 2 | 654 | 87 | ||
GenBank mRNA | AK119137.1 | - | 1 - 2 | 550 | 87 | |
AY084590.1 | - | 1 - 2 | 611 | 87 | ||
BT005382.1 | - | 1 - 2 | 528 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |