Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BDG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334171.1 | NP_001323396.1 | 2 - 3 | 1064 | 68 | |
NM_001334172.1 | NP_001323395.1 | 2 - 3 | 1022 | 68 | ||
NM_105142.4 | NP_564837.1 | 2 - 3 | 990 | 68 | ||
GenBank mRNA | AK119137.1 | - | 2 - 3 | 886 | 68 | |
AY084590.1 | - | 2 - 3 | 947 | 68 | ||
BT005382.1 | - | 2 - 3 | 864 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |