Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334199.1 | NP_001322349.1 | 9 - 10 | 1201 | 83 | |
NM_105199.3 | NP_564846.1 | 9 - 10 | 1046 | 83 | ||
GenBank mRNA | AY045845.1 | - | 9 - 10 | 814 | 83 | |
AY117153.1 | - | 9 - 10 | 805 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |