Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_110977.3 | NP_186761.1 | 8 - 9 | 1638 | 69 | |
GenBank mRNA | AY094438.1 | - | 8 - 9 | 1376 | 69 | |
BT002753.1 | - | 8 - 9 | 1344 | 69 | ||
U83500.1 | - | 8 - 9 | 1364 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |