Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202860.2 | NP_001189789.1 | 19 - 20 | 4126 | 143 | |
NM_110998.5 | NP_186781.4 | 19 - 20 | 4166 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |