Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203778.1 | NP_001190707.1 | 3 - 4 | 531 | 71 | |
NM_001340798.1 | NP_001329574.1 | 3 - 4 | 584 | 71 | ||
NM_001340799.1 | NP_001329573.1 | 1 - 2 | 150 | 71 | ||
NM_117341.5 | NP_193008.1 | 4 - 5 | 441 | 71 | ||
NM_179036.4 | NP_849367.1 | 3 - 4 | 607 | 71 | ||
NM_179037.3 | NP_849368.1 | 3 - 4 | 279 | 71 | ||
GenBank mRNA | AF325104.1 | - | 2 - 3 | 207 | 71 | |
AF370209.1 | - | 3 - 4 | 265 | 71 | ||
AY056344.1 | - | 2 - 3 | 207 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |