Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203778.1 | NP_001190707.1 | 4 - 5 | 675 | 98 | |
NM_001340798.1 | NP_001329574.1 | 4 - 5 | 728 | 98 | ||
NM_001340799.1 | NP_001329573.1 | 2 - 3 | 294 | 98 | ||
NM_117341.5 | NP_193008.1 | 5 - 6 | 585 | 98 | ||
NM_179036.4 | NP_849367.1 | 4 - 5 | 751 | 98 | ||
NM_179037.3 | NP_849368.1 | 4 - 5 | 423 | 98 | ||
GenBank mRNA | AF325104.1 | - | 3 - 4 | 351 | 98 | |
AF370209.1 | - | 4 - 5 | 409 | 98 | ||
AY056344.1 | - | 3 - 4 | 351 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |