Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MHK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036545.1 | NP_001031622.1 | 12 - 13 | 1229 | 136 | |
NM_001125502.1 | NP_001118974.1 | 14 - 15 | 1513 | 136 | ||
NM_001203783.2 | NP_001190712.1 | 14 - 15 | 1550 | 136 | ||
NM_001340823.1 | NP_001329480.1 | 14 - 15 | 1523 | 136 | ||
NM_117371.1 | NP_193038.1 | 12 - 13 | 1229 | 136 | ||
NM_179039.4 | NP_849370.1 | 14 - 15 | 1635 | 136 | ||
GenBank mRNA | AF360324.1 | - | 14 - 15 | 1484 | 136 | |
AY056332.1 | - | 12 - 13 | 1016 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |