Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MHK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036545.1 | NP_001031622.1 | 2 - 3 | 371 | 132 | |
NM_001125502.1 | NP_001118974.1 | 4 - 5 | 655 | 132 | ||
NM_001340823.1 | NP_001329480.1 | 4 - 5 | 665 | 132 | ||
NM_001340824.1 | NP_001329479.1 | 4 - 5 | 665 | 132 | ||
NM_117371.1 | NP_193038.1 | 2 - 3 | 371 | 132 | ||
NM_179039.4 | NP_849370.1 | 4 - 5 | 777 | 132 | ||
GenBank mRNA | AF360324.1 | - | 4 - 5 | 626 | 132 | |
AY056332.1 | - | 2 - 3 | 158 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |